The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037713.4 | NP_001032802.2 | 1 - 2 | 84 | 67 |
NM_001291153.1 | NP_001278082.1 | 1 - 2 | 84 | 67 | |
GenBank mRNA | AK046736.1 | - | 1 - 2 | 51 | 67 |
AK140315.1 | - | 1 - 2 | 49 | 67 | |
BY726623.1 | - | 1 - 2 | 84 | 67 | |
EL957551.1 | - | 1 - 2 | 74 | 67 |