Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snph
Entrez Gene ID
241727
Gene Name
syntaphilin
Gene Aliases
6430515A01, AW045671, AW556958, AW743098, mKIAA0374
Chromosome Location
Chr.2: 151590549 - 151633030 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291077.1 NP_001278006.1 3 - 4 346 90
GenBank mRNA BC096541.1 - 3 - 4 238 90
BC117897.1 - 3 - 4 272 90

View Full Product Details