The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040403.1 | NP_001035493.1 | 1 - 2 | 174 | 79 |
NM_001284227.1 | NP_001271156.1 | 1 - 2 | 174 | 79 | |
NM_008028.2 | NP_032054.1 | 1 - 2 | 174 | 79 | |
GenBank mRNA | AK004589.1 | - | 1 - 2 | 112 | 79 |
AK030199.1 | - | 1 - 2 | 137 | 79 | |
AK149530.1 | - | 1 - 2 | 112 | 79 | |
AK170557.1 | - | 1 - 2 | 175 | 79 | |
AK187814.1 | - | 1 - 2 | 58 | 79 | |
AK200150.1 | - | 1 - 2 | 85 | 79 | |
AK200750.1 | - | 1 - 2 | 85 | 79 | |
BC070423.1 | - | 1 - 2 | 132 | 79 | |
BY195482.1 | - | 1 - 2 | 175 | 79 | |
BY204171.1 | - | 1 - 2 | 175 | 79 |