Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Raly
Entrez Gene ID
19383
Gene Name
hnRNP-associated with lethal yellow
Gene Aliases
AI663842, Merc
Chromosome Location
Chr.2: 154791097 - 154871010 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001139513.1 NP_001132985.1 4 - 5 823 80

View Full Product Details