The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013865.2 | NP_038893.1 | 3 - 4 | 244 | 63 |
NM_180956.1 | NP_851287.1 | 3 - 4 | 244 | 63 | |
GenBank mRNA | AB033922.1 | - | 3 - 4 | 158 | 63 |
AK002272.1 | - | 2 - 3 | 158 | 63 | |
AK004689.1 | - | 3 - 4 | 190 | 63 | |
AK036277.1 | - | 3 - 4 | 169 | 63 | |
AK037487.1 | - | 3 - 4 | 170 | 63 | |
AK154927.1 | - | 3 - 4 | 211 | 63 | |
BC018504.1 | - | 3 - 4 | 178 | 63 | |
BY125191.1 | - | 3 - 4 | 245 | 63 |