Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acox1
Entrez Gene ID
11430
Gene Name
acyl-Coenzyme A oxidase 1, palmitoyl
Gene Aliases
AOX, Acox, D130055E20Rik, Paox
Chromosome Location
Chr.11: 116171883 - 116199045 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271898.1 NP_001258827.1 11 - 12 106
NM_015729.3 NP_056544.2 11 - 12 106
GenBank mRNA AB034914.1 - 11 - 12 106
AF006688.1 - 11 - 12 106
AK034760.1 - 7 - 8 106
AK040566.1 - 11 - 12 106
AK128941.1 - 11 - 12 1626 106
BC054727.1 - 11 - 12 106
BC056448.1 - 11 - 12 106

View Full Product Details