The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048145.1 | NP_001041610.1 | 8 - 9 | 538 | 77 |
NM_001286494.1 | NP_001273423.1 | 7 - 8 | 475 | 77 | |
NM_029139.1 | NP_083415.1 | 9 - 10 | 602 | 77 | |
GenBank mRNA | AK015606.1 | - | 9 - 10 | 598 | 77 |
AY899935.1 | - | 8 - 9 | 538 | 77 | |
AY899936.1 | - | 9 - 10 | 602 | 77 | |
BC100478.1 | - | 7 - 8 | 456 | 77 | |
BC141326.1 | - | 8 - 9 | 538 | 77 | |
BC141347.1 | - | 8 - 9 | 538 | 77 |