Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dram1
- Entrez Gene ID
-
71712
- Gene Name
- DNA-damage regulated autophagy modulator 1
- Gene Aliases
-
1200002N14Rik, Dram
- Chromosome Location
-
Chr.10: 88322801 - 88364383 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC021433.1 |
- |
2 - 3 |
316 |
126 |
View Full Product Details