Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gosr2
- Entrez Gene ID
-
56494
- Gene Name
- golgi SNAP receptor complex member 2
- Gene Aliases
-
2310032N09Rik, C76855, Gs27, SNARE, membrin
- Chromosome Location
-
Chr.11: 103676849 - 103697738 on Build GRCm38
- Gene Symbol
- Lgmn
- Entrez Gene ID
-
19141
- Gene Name
- legumain
- Gene Aliases
-
AEP, AI746452, AU022324, Prsc1
- Chromosome Location
-
Chr.12: 102394098 - 102439697 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK180466.1 |
- |
- |
|
66 |
View Full Product Details