Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Elk3
Entrez Gene ID
13713
Gene Name
ELK3, member of ETS oncogene family
Gene Aliases
D430049E23Rik, Erp, Etrp, Net, Sap-2
Chromosome Location
Chr.10: 93247415 - 93311167 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282967.1 NP_001269896.1 1 - 2 408 62
NM_013508.2 NP_038536.2 1 - 2 408 62
GenBank mRNA AK052546.1 - 1 - 2 85 62
AK088129.1 - 1 - 2 223 62
AK133847.1 - 1 - 2 104 62
BC005686.1 - 1 - 2 390 62
BC053402.1 - 1 - 2 159 62
BC055735.1 - 1 - 2 181 62
L19953.1 - 1 - 2 299 62
S82864.1 - 1 - 2 91 62
Z32815.1 - 1 - 2 292 62

View Full Product Details