Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lin7a
Entrez Gene ID
108030
Gene Name
lin-7 homolog A (C. elegans)
Gene Aliases
AI848705, LIN-7A, MALS-1, TIP-33, Veli, Veli1
Chromosome Location
Chr.10: 107271798 - 107425146 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284329.1 NP_001271258.1 1 - 2 870 97
GenBank mRNA AK077191.1 - 1 - 2 870 97

View Full Product Details