Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krr1
Entrez Gene ID
52705
Gene Name
KRR1, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases
2610511F02Rik, AI255219, AI428520, D10Ertd773e, Hrb2
Chromosome Location
Chr.10: 111972695 - 111988430 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178610.4 NP_848725.2 5 - 6 697 114
GenBank mRNA AK011972.1 - 5 - 6 607 114
AK012108.1 - 5 - 6 635 114
AK030560.1 - 5 - 6 609 114
AK032472.1 - 5 - 6 609 114
AK036447.1 - 4 - 5 695 114
AK041854.1 - 5 - 6 609 114
AK146926.1 - 5 - 6 615 114
AK168675.1 - 5 - 6 609 114
BC089510.1 - 5 - 6 614 114
BC094236.1 - 5 - 6 598 114

View Full Product Details