Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Best3
Entrez Gene ID
382427
Gene Name
bestrophin 3
Gene Aliases
Vmd2l3, mBest4
Chromosome Location
Chr.10: 116986314 - 117025687 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007583.1 NP_001007584.1 8 - 9 1462 74
GenBank mRNA AY450426.1 - 8 - 9 1462 74

View Full Product Details