Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fendrr
Entrez Gene ID
68790
Gene Name
Foxf1 adjacent non-coding developmental regulatory RNA
Gene Aliases
1110050K14Rik, LL66, linc-Foxf1a, lincFoxf1
Chromosome Location
Chr.8: 121054882 - 121083110 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK087264.1 - 1 - 2 60 70
JQ973641.2 - 1 - 2 137 70


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB664441.1 - 1 - 2 60 Chr. 8 - 121065746 - 121083032 [-] on Build GRCm38 70


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream predicted gene, 26878 Gm26878 -192.3kb Anti-sense Chr. - 120879399 - 120881217 on Build GRCm38
Downstream forkhead box F1 Foxf1 AI450827, FREAC1, Foxf1a, Freac-1, HFH-8, Hfh8 1.4kb Anti-sense Chr. - 121084386 - 121088154 on Build GRCm38

View Full Product Details