The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027919.4 | NP_082195.2 | 4 - 5 | 716 | 73 |
GenBank mRNA | AK005036.2 | - | 2 - 3 | 1566 | 73 |
AY219871.1 | - | 4 - 5 | 716 | 73 | |
AY219872.1 | - | 3 - 4 | 500 | 73 | |
BC055825.1 | - | 4 - 5 | 649 | 73 |