The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027919.4 | NP_082195.2 | 6 - 7 | 1050 | 72 |
GenBank mRNA | AY219871.1 | - | 6 - 7 | 1050 | 72 |
AY219872.1 | - | 5 - 6 | 834 | 72 | |
BC055825.1 | - | 6 - 7 | 983 | 72 | |
BC066149.1 | - | 2 - 3 | 347 | 72 |