Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sin3b
Entrez Gene ID
20467
Gene Name
transcriptional regulator, SIN3B (yeast)
Gene Aliases
2810430C10Rik
Chromosome Location
Chr.8: 72723270 - 72758203 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009188.4 NP_033214.2 8 - 9 1064 70
GenBank mRNA AK087970.1 - 1 - 2 161 70
AK159076.1 - 8 - 9 1047 70
AK173005.1 - 7 - 8 3014 70
BC129859.1 - 8 - 9 1036 70
L38622.2 - 8 - 9 1036 70

View Full Product Details