The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133850.2 | NP_598611.2 | 1 - 2 | 182 | 70 |
GenBank mRNA | AK008447.1 | - | 1 - 2 | 137 | 70 |
AK009274.1 | - | 1 - 2 | 157 | 70 | |
AK046582.1 | - | 1 - 2 | 132 | 70 | |
AK077349.1 | - | 1 - 2 | 123 | 70 | |
AK078774.1 | - | 1 - 2 | 141 | 70 | |
AK089926.1 | - | 1 - 2 | 125 | 70 | |
AK164805.1 | - | 1 - 2 | 91 | 70 | |
BC003310.1 | - | 1 - 2 | 115 | 70 | |
BC065794.1 | - | 1 - 2 | 83 | 70 | |
BC092215.1 | - | 1 - 2 | 99 | 70 | |
BC094216.1 | - | 1 - 2 | 139 | 70 |