The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016905.2 | NP_058601.2 | 7 - 8 | 1147 | 78 |
GenBank mRNA | AB027012.1 | - | 7 - 8 | 1103 | 78 |
AK010340.1 | - | 7 - 8 | 1144 | 78 | |
AK011103.1 | - | 7 - 8 | 1148 | 78 | |
AK045694.1 | - | 7 - 8 | 1121 | 78 | |
AK088151.1 | - | 7 - 8 | 1148 | 78 | |
AK168716.1 | - | 7 - 8 | 1107 | 78 | |
BC016602.1 | - | 7 - 8 | 1147 | 78 | |
BC050151.1 | - | 7 - 8 | 1118 | 78 |