Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Snph
- Entrez Gene ID
-
241727
- Gene Name
- syntaphilin
- Gene Aliases
-
6430515A01, AW045671, AW556958, AW743098, mKIAA0374
- Chromosome Location
-
Chr.2: 151590549 - 151633030 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK047595.1 |
- |
2 - 3 |
226 |
149 |
View Full Product Details