Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Endov
Entrez Gene ID
338371
Gene Name
endonuclease V
Gene Aliases
A730011L01Rik
Chromosome Location
Chr.11: 119491323 - 119511465 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164636.1 NP_001158108.1 4 - 5 456 81
NM_177394.3 NP_796368.1 3 - 4 508 81
GenBank mRNA AK042626.1 - 3 - 4 396 81
AK084233.1 - 4 - 5 457 81
AK084548.1 - 4 - 5 332 81
AK170595.1 - 3 - 4 501 81
AK172181.1 - 3 - 4 475 81
AK179832.1 - 2 - 3 153 81
BC056486.1 - 3 - 4 428 81

View Full Product Details