Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Brca1
Entrez Gene ID
12189
Gene Name
breast cancer 1
Gene Aliases
-
Chromosome Location
Chr.11: 101488754 - 101551956 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009764.3 NP_033894.3 11 - 12 4262 72
GenBank mRNA AB221610.1 - 2 - 3 1491 72
BC068303.1 - 11 - 12 4098 72
U31625.1 - 11 - 12 4119 72
U32446.1 - 11 - 12 4172 72
U36475.1 - 10 - 11 4082 72
U68174.1 - 11 - 12 4166 72

View Full Product Details