Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Brca1
Entrez Gene ID
12189
Gene Name
breast cancer 1
Gene Aliases
-
Chromosome Location
Chr.11: 101488754 - 101551956 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009764.3 NP_033894.3 22 - 23 5500 75
GenBank mRNA AB221610.1 - 13 - 14 2729 75
BC068303.1 - 22 - 23 5333 75
U31625.1 - 22 - 23 5357 75
U32446.1 - 22 - 23 5410 75
U35641.1 - 22 - 23 5393 75
U36475.1 - 21 - 22 5320 75
U68174.1 - 22 - 23 5404 75

View Full Product Details