Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arhgap40
Entrez Gene ID
545481
Gene Name
Rho GTPase activating protein 40
Gene Aliases
C20orf95, Gm14203, OTTMUSG00000015933
Chromosome Location
Chr.2: 158512796 - 158550628 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145015.1 NP_001138487.1 5 - 6 790 128

View Full Product Details