Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myh7b
Entrez Gene ID
668940
Gene Name
myosin, heavy chain 7B, cardiac muscle, beta
Gene Aliases
Myh14
Chromosome Location
Chr.2: 155611243 - 155634307 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085378.2 NP_001078847.1 11 - 12 994 73

View Full Product Details