Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Edem2
Entrez Gene ID
108687
Gene Name
ER degradation enhancer, mannosidase alpha-like 2
Gene Aliases
9530090G24Rik, AI327354
Chromosome Location
Chr.2: 155701673 - 155729475 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145537.2 NP_663512.2 4 - 5 503 84
GenBank mRNA AK029164.1 - 4 - 5 454 84
AK075650.1 - 4 - 5 417 84
AK079296.1 - 4 - 5 439 84
AK080636.1 - 4 - 5 448 84
AK149892.1 - 4 - 5 439 84
AK155024.1 - 4 - 5 466 84
AK166998.1 - 4 - 5 439 84
BC008268.1 - 4 - 5 414 84

View Full Product Details