Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acsf3
Entrez Gene ID
257633
Gene Name
acyl-CoA synthetase family member 3
Gene Aliases
BB101783
Chromosome Location
Chr.8: 122775493 - 122817882 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK219530.1 - 1 - 2 49 60

View Full Product Details