Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmo1
Entrez Gene ID
327956
Gene Name
vitelline membrane outer layer 1 homolog (chicken)
Gene Aliases
Gm741
Chromosome Location
Chr.11: 70513516 - 70514616 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013607.1 NP_001013625.1 2 - 3 323 86

View Full Product Details