Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Alox15
Entrez Gene ID
11687
Gene Name
arachidonate 15-lipoxygenase
Gene Aliases
12-LO, 12/15-LO, 15-LOX, Alox12l, L-12LO
Chromosome Location
Chr.11: 70344147 - 70375918 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009660.3 NP_033790.3 5 - 6 671 107
GenBank mRNA AK142371.1 - 5 - 6 672 107
BC056625.1 - 5 - 6 658 107
BC081546.1 - 5 - 6 657 107
CT010263.1 - 5 - 6 645 107
L34570.1 - 5 - 6 655 107
U04331.1 - 5 - 6 645 107

View Full Product Details