The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271358.1 | NP_001258287.1 | 10 - 11 | 932 | 75 |
NM_001271359.1 | NP_001258288.1 | 10 - 11 | 932 | 75 | |
NM_001271360.1 | NP_001258289.1 | 10 - 11 | 932 | 75 | |
NM_145429.5 | NP_663404.1 | 10 - 11 | 932 | 75 | |
GenBank mRNA | AK089542.1 | - | 10 - 11 | 868 | 75 |
AK154874.1 | - | 10 - 11 | 852 | 75 | |
AK159317.1 | - | 10 - 11 | 860 | 75 | |
AK162889.1 | - | 10 - 11 | 869 | 75 | |
AK170647.1 | - | 10 - 11 | 879 | 75 | |
AK170889.1 | - | 10 - 11 | 842 | 75 | |
BC016642.1 | - | 10 - 11 | 814 | 75 |