The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021491.3 | NP_067466.1 | 8 - 9 | 2335 | 93 |
GenBank mRNA | AJ250461.1 | - | 6 - 7 | 1866 | 93 |
AK019476.2 | - | 9 - 10 | 2387 | 93 | |
AK076416.1 | - | 8 - 9 | 2312 | 93 | |
AK139226.1 | - | 8 - 9 | 2311 | 93 | |
AK140788.1 | - | 4 - 5 | 337 | 93 | |
BC043077.1 | - | 8 - 9 | 2335 | 93 | |
BC046980.1 | - | 7 - 8 | 2155 | 93 |