The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175935.3 | NP_787949.2 | 5 - 6 | 902 | 75 |
GenBank mRNA | AK002966.1 | - | 6 - 7 | 765 | 75 |
AK165395.1 | - | 5 - 6 | 756 | 75 | |
AY186239.1 | - | 5 - 6 | 748 | 75 | |
BC069959.1 | - | 5 - 6 | 902 | 75 |