Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmub2
Entrez Gene ID
72053
Gene Name
transmembrane and ubiquitin-like domain containing 2
Gene Aliases
2010008E23Rik
Chromosome Location
Chr.11: 102284820 - 102289427 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK008158.1 - 1 - 2 78 75
AK132118.1 - 1 - 2 122 75

View Full Product Details