Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Manbal
Entrez Gene ID
69161
Gene Name
mannosidase, beta A, lysosomal-like
Gene Aliases
1810024K12Rik, AI836500
Chromosome Location
Chr.2: 157367594 - 157396763 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK169993.1 - 2 - 3 239 59
BC030375.1 - 2 - 3 233 59

View Full Product Details