The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025853.3 | NP_080129.2 | 8 - 9 | 822 | 77 |
GenBank mRNA | AK006246.1 | - | 8 - 9 | 823 | 77 |
AK017740.1 | - | 8 - 9 | 823 | 77 | |
AK017813.1 | - | 8 - 9 | 777 | 77 | |
AK139887.1 | - | 8 - 9 | 1392 | 77 | |
AK145373.1 | - | 9 - 10 | 650 | 77 | |
BC025079.1 | - | 8 - 9 | 805 | 77 | |
BC046807.1 | - | 8 - 9 | 783 | 77 |