Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc16a13
Entrez Gene ID
69309
Gene Name
solute carrier family 16 (monocarboxylic acid transporters), member 13
Gene Aliases
1700007D07Rik
Chromosome Location
Chr.11: 70216789 - 70222794 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172371.3 NP_758959.1 2 - 3 658 75
GenBank mRNA AK028143.1 - 2 - 3 643 75
AK028759.1 - 2 - 3 659 75
BC039949.1 - 2 - 3 446 75

View Full Product Details