The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013380.2 | NP_001013398.2 | 2 - 3 | 430 | 81 |
GenBank mRNA | AK050575.1 | - | 2 - 3 | 399 | 81 |
AK140743.1 | - | 2 - 3 | 198 | 81 | |
AK147899.1 | - | 2 - 3 | 197 | 81 | |
AK152157.1 | - | 2 - 3 | 195 | 81 | |
AK161434.1 | - | 2 - 3 | 292 | 81 | |
AK161494.1 | - | 2 - 3 | 292 | 81 | |
BC058645.1 | - | 2 - 3 | 388 | 81 |