The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027946.3 | NP_082222.1 | 1 - 2 | 392 | 93 |
GenBank mRNA | AK005902.1 | - | 1 - 2 | 386 | 93 |
AK047435.1 | - | 1 - 2 | 394 | 93 | |
AK080823.1 | - | 1 - 2 | 365 | 93 | |
AK154757.1 | - | 1 - 2 | 383 | 93 | |
AK165674.1 | - | 1 - 2 | 383 | 93 | |
BB646282.1 | - | 1 - 2 | 393 | 93 | |
BC048165.1 | - | 1 - 2 | 308 | 93 | |
BC048722.1 | - | 1 - 2 | 311 | 93 |