Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gdap1l1
Entrez Gene ID
228858
Gene Name
ganglioside-induced differentiation-associated protein 1-like 1
Gene Aliases
Gdap1l
Chromosome Location
Chr.2: 163438419 - 163455324 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144891.2 NP_659140.2 4 - 5 726 63
GenBank mRNA AK140488.1 - 4 - 5 717 63
AK141371.1 - 4 - 5 718 63
BC019941.1 - 4 - 5 740 63

View Full Product Details