The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009036.1 | NP_033062.1 | 4 - 5 | 426 | 80 |
GenBank mRNA | AK052449.1 | - | 4 - 5 | 1552 | 80 |
AK131814.1 | - | 4 - 5 | 407 | 80 | |
AK131832.1 | - | 4 - 5 | 412 | 80 | |
BC152311.1 | - | 4 - 5 | 420 | 80 | |
Y10926.1 | - | 4 - 5 | 426 | 80 |