Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Svs2
Entrez Gene ID
53878
Gene Name
seminal vesicle secretory protein 2
Gene Aliases
BB115391, Semg1, SvsII
Chromosome Location
Chr.2: 164235929 - 164238341 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017390.4 NP_059086.3 2 - 3 1267 127
GenBank mRNA AK020661.1 - 2 - 3 711 127
AK035204.1 - 2 - 3 1192 127
AK040724.1 - 2 - 3 1193 127
AK090151.1 - 2 - 3 1189 127
AK090302.1 - 2 - 3 1193 127
AK144285.1 - 2 - 3 1192 127
AK144389.1 - 2 - 3 1192 127
AK144429.1 - 2 - 3 1192 127
AK144524.1 - 2 - 3 1189 127
AK165704.1 - 2 - 3 1192 127
AK165713.1 - 2 - 3 1192 127
AK165727.1 - 2 - 3 1189 127
AK165753.1 - 2 - 3 1192 127
AK165825.1 - 2 - 3 1192 127
BC107275.1 - 2 - 3 1192 127
BC107276.1 - 2 - 3 1191 127

View Full Product Details