The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018883.2 | NP_061371.2 | 15 - 16 | 1607 | 61 |
GenBank mRNA | AF117384.1 | - | 15 - 16 | 1562 | 61 |
AK144483.1 | - | 14 - 15 | 1877 | 61 | |
BC017529.1 | - | 15 - 16 | 1572 | 61 |