Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asb16
Entrez Gene ID
217217
Gene Name
ankyrin repeat and SOCS box-containing 16
Gene Aliases
-
Chromosome Location
Chr.11: 102268823 - 102278061 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148953.2 NP_683755.1 1 - 2 300 77
GenBank mRNA AF403044.1 - 1 - 2 300 77
BC127245.1 - 1 - 2 296 77
BC127246.1 - 1 - 2 299 77
BC152348.1 - 1 - 2 300 77
BC152349.1 - 1 - 2 300 77

View Full Product Details