The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010970.1 | NP_035100.1 | 1 - 1 | 551 | 147 |
GenBank mRNA | X89688.1 | - | 1 - 1 | 177 | 147 |