The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001131020.1 | NP_001124492.1 | 3 - 4 | 695 | 64 |
NM_010277.3 | NP_034407.2 | 3 - 4 | 695 | 64 | |
GenBank mRNA | AF332061.1 | - | 3 - 4 | 522 | 64 |
AF332062.1 | - | 3 - 4 | 522 | 64 | |
AK079577.1 | - | 3 - 4 | 629 | 64 | |
AK140151.1 | - | 3 - 4 | 627 | 64 | |
BC100728.1 | - | 3 - 4 | 574 | 64 | |
BC100737.1 | - | 3 - 4 | 567 | 64 | |
BC101968.1 | - | 3 - 4 | 574 | 64 | |
BC103571.1 | - | 3 - 4 | 568 | 64 | |
BC139357.1 | - | 3 - 4 | 695 | 64 | |
BC139358.1 | - | 3 - 4 | 695 | 64 | |
DQ887822.1 | - | 1 - 2 | 110 | 64 | |
EF101554.1 | - | 3 - 4 | 341 | 64 |