Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Urb2
Entrez Gene ID
382038
Gene Name
URB2 ribosome biogenesis 2 homolog (S. cerevisiae)
Gene Aliases
C77575, mKIAA0133
Chromosome Location
Chr.8: 124021442 - 124048505 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029876.1 NP_001025047.1 5 - 6 3913 122
GenBank mRNA AK122209.1 - 3 - 4 3550 122
AK133906.1 - 6 - 7 4214 122
BC096020.1 - 3 - 4 2296 122
BC151125.1 - 5 - 6 3992 122

View Full Product Details