The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026197.3 | NP_080473.1 | 7 - 8 | 914 | 68 |
GenBank mRNA | AK011780.1 | - | 7 - 8 | 915 | 68 |
AK012739.1 | - | 7 - 8 | 915 | 68 | |
AK013306.1 | - | 7 - 8 | 926 | 68 | |
AK029045.1 | - | 7 - 8 | 897 | 68 | |
AK145662.1 | - | 7 - 8 | 914 | 68 | |
AK168798.1 | - | 7 - 8 | 864 | 68 | |
BC003467.1 | - | 7 - 8 | 886 | 68 | |
BC022902.1 | - | 8 - 9 | 1047 | 68 | |
BC037452.1 | - | 9 - 10 | 1059 | 68 | |
BC049897.1 | - | 7 - 8 | 843 | 68 | |
BC054724.1 | - | 7 - 8 | 850 | 68 |