The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023709.4 | NP_076198.2 | 4 - 5 | 571 | 60 |
GenBank mRNA | AK144514.1 | - | 4 - 5 | 572 | 60 |
BC058748.1 | - | 4 - 5 | 549 | 60 | |
U89513.1 | - | 4 - 5 | 532 | 60 |