Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc13
Entrez Gene ID
234875
Gene Name
tetratricopeptide repeat domain 13
Gene Aliases
BC017545
Chromosome Location
Chr.8: 124671327 - 124722013 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145607.3 NP_663582.2 12 - 13 1485 162
GenBank mRNA AK144842.1 - 12 - 13 1486 162
AK148102.1 - 14 - 15 1715 162
AK160894.1 - 14 - 15 1723 162
BC017545.1 - 6 - 7 699 162
BC072577.1 - 12 - 13 1478 162
BC079851.1 - 13 - 14 1540 162

View Full Product Details