Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ribc1
Entrez Gene ID
66611
Gene Name
RIB43A domain with coiled-coils 1
Gene Aliases
2610028I09Rik, W08639
Chromosome Location
Chr.X: 152004580 - 152018219 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025660.2 NP_079936.1 4 - 5 426 92
GenBank mRNA AK011592.1 - 4 - 5 426 92
AK017461.1 - 4 - 5 318 92
BC050755.1 - 4 - 5 414 92

View Full Product Details